Assay Details
| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032397.2 | NP_115773.2 | 6 - 7 | 1323 | 65 | |
| GenBank mRNA | AF116525.1 | - | 5 - 6 | 1351 | 65 | |
| AF357239.1 | - | 6 - 7 | 1279 | 65 | ||
| AK041033.1 | - | 6 - 7 | 1324 | 65 | ||
| AK043611.1 | - | 6 - 7 | 1293 | 65 | ||
| AY258143.1 | - | 5 - 6 | 1162 | 65 | ||
| BC090622.1 | - | 7 - 8 | 1666 | 6832359 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn1
Genomic Map