Assay Details
| Gene Symbol: Copb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033370.3 | NP_203534.1 | 7 - 8 | 976 | 114 | |
| GenBank mRNA | AF231925.1 | - | 6 - 7 | 915 | 114 | |
| AK011453.1 | - | 7 - 8 | 1000 | 114 | ||
| AK135168.1 | - | 7 - 8 | 1008 | 114 | ||
| AK149828.1 | - | 7 - 8 | 994 | 114 | ||
| AK152222.1 | - | 7 - 8 | 1003 | 114 | ||
| AK161544.1 | - | 7 - 8 | 1003 | 114 | ||
| AK172214.1 | - | 7 - 8 | 978 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copb1
Genomic Map