Assay Details
| Gene Symbol: Copb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033370.3 | NP_203534.1 | 10 - 11 | 1348 | 73 | |
| GenBank mRNA | AF231925.1 | - | 9 - 10 | 1287 | 73 | |
| AK011453.1 | - | 10 - 11 | 1372 | 73 | ||
| AK135168.1 | - | 10 - 11 | 1380 | 73 | ||
| AK149828.1 | - | 10 - 11 | 1366 | 73 | ||
| AK152222.1 | - | 10 - 11 | 1375 | 73 | ||
| AK161544.1 | - | 10 - 11 | 1375 | 73 | ||
| AK172214.1 | - | 10 - 11 | 1350 | 73 | ||
| BC030837.1 | - | 10 - 11 | 1348 | 4022216 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Copb1
Genomic Map