Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173372.1 | NP_001166843.1 | 8 - 9 | 360 | 84 | |
| NM_033648.1 | NP_387468.1 | 8 - 9 | 363 | 84 | ||
| GenBank mRNA | AK018728.1 | - | 8 - 9 | 360 | 84 | |
| AU041511.1 | - | 1 - 2 | 84 | |||
| BC086918.1 | - | 9 - 10 | 624 | 6762406 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520530 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136212 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24784135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136212 | - | - |
| Whole Mouse Genome G4122A | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | AK018728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | AU041511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | BC086918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | NM_001173372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P141926 | NM_033648.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136212 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05601541 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05617217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05617222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05617218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05601540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24784120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05733725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05046160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05617224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05706888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05617221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05104075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05799561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05372422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05326894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05349626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05104082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05570415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05777265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05349621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05439486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24784116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547213 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547214 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547215 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547216 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418207_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547206 | - | - |
| Mouse Genome 430 2.0 Array | 1418207_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418207_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108017_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108017_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600022985.mm.1 | TC0600002992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600022985.mm.1 | NM_001173372.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022985.mm.1 | NM_001173372.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022985.mm.1 | NM_033648.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022985.mm.1 | NM_033648.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022983.mm.1 | TC0600002992.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600022983.mm.1 | NM_001173372.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022983.mm.1 | NM_001173372.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022983.mm.1 | NM_033648.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022983.mm.1 | NM_033648.1 | Refseq |
| Mouse Expression Set 430 | 1418207_PM_at | - | - |
| Mouse Expression Set 430 | 1418207_at | - | - |
| Murine Genome U74v2 Set | 171114_PM_at | - | - |
| Murine Genome U74v2 Set | 171114_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418207_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418207_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418207_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418207_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 704669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5270292 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270292 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2720117 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670751 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2720117 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670751 | - | - |
| mouse-6 v1.1 (Illumina) | scl28501.8.1_119-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001091.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370725 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270292 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28501.8.1_119-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001091.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215552 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670751 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670751 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215552 | - | - |

Fxyd4
Genomic Map