Assay Details
| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054084.2 | NP_473425.2 | 3 - 4 | 318 | 63 | |
| GenBank mRNA | AK136518.1 | - | 3 - 4 | 312 | 63 | |
| BC111865.1 | - | 3 - 4 | 318 | 40049329 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Calcb
Genomic Map