Assay Details
| Gene Symbol: Calcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054084.2 | NP_473425.2 | 4 - 5 | 506 | 98 | |
| GenBank mRNA | AI324836.1 | - | 1 - 2 | 98 | ||
| AK136518.1 | - | 4 - 5 | 500 | 98 | ||
| BC111865.1 | - | 4 - 5 | 506 | 40049329 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709903 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103407 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25437412 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103407 | - | - |
| Whole Mouse Genome G4122A | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406157 | AI324836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406157 | NM_054084.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136934 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136500 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25437360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05977885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05977887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06466074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06041900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06293305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06359579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05977889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05934055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06041901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06476926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06476922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06545073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06293296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05995902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25437428 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10556551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556550 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422639_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10556546 | - | - |
| Mouse Genome 430 2.0 Array | 1422639_at | - | - |
| Mouse Genome 430 2.0 Array | 1422639_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116903_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116903_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700013936.mm.1 | TC0700001672.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700013936.mm.1 | NM_054084.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700013936.mm.1 | NM_054084.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700013935.mm.1 | TC0700001672.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700013935.mm.1 | NM_054084.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700013935.mm.1 | NM_054084.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700013934.mm.1 | TC0700001672.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700013934.mm.1 | NM_054084.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700013934.mm.1 | NM_054084.2 | Refseq |
| Mouse Expression Set 430 | 1422639_PM_at | - | - |
| Mouse Expression Set 430 | 1422639_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422639_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422639_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422639_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422639_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 569420 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2690450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2690450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002690450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252436 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252436 | - | - |
| mouse-6 v1.1 (Illumina) | scl32159.5.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2690450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32159.5.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2690450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252436 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252436 | - | - |

Calcb
Genomic Map