Assay Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080844.4 | NP_543120.1 | 3 - 4 | 617 | 71 | |
| GenBank mRNA | AK050295.1 | - | 2 - 3 | 459 | 71 | |
| BC011165.1 | - | 2 - 3 | 424 | 71 | ||
| BC019447.1 | - | 3 - 4 | 577 | 5125534 | 71 | |
| BC033377.1 | - | 2 - 3 | 461 | 5123932 | 71 | |
| DV662863.1 | - | 3 - 4 | 590 | 71 | ||
| S47225.1 | - | 2 - 3 | 420 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinc1
Genomic Map