Assay Details
| Gene Symbol: Tm9sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133352.2 | NP_579930.1 | 12 - 13 | 1682 | 68 | |
| GenBank mRNA | AF269151.1 | - | 12 - 13 | 1614 | 68 | |
| AK075807.1 | - | 12 - 13 | 1648 | 68 | ||
| AK147380.1 | - | 12 - 13 | 1677 | 68 | ||
| AK147629.1 | - | 12 - 13 | 1676 | 68 | ||
| AK220279.1 | - | 12 - 13 | 1658 | 68 | ||
| BC004799.1 | - | 11 - 12 | 1311 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf3
Genomic Map