Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 1 - 2 | 111 | 101 | |
| GenBank mRNA | AK085754.1 | - | 1 - 2 | 96 | 101 | |
| AK142599.1 | - | 101 | 101 | |||
| AK146549.1 | - | 1 - 2 | 96 | 101 | ||
| AK149559.1 | - | 1 - 2 | 96 | 101 | ||
| AK149595.1 | - | 1 - 2 | 96 | 101 | ||
| AK168405.1 | - | 1 - 2 | 96 | 101 | ||
| AK168419.1 | - | 1 - 2 | 96 | 101 | ||
| AK168938.1 | - | 1 - 2 | 96 | 101 | ||
| AK169187.1 | - | 1 - 2 | 96 | 101 | ||
| BC012313.1 | - | 1 - 2 | 111 | 3981648 | 101 | |
| BC020295.1 | - | 1 - 2 | 80 | 101 | ||
| BC022986.1 | - | 1 - 2 | 89 | 5123741 | 101 | |
| BC058216.1 | - | 1 - 2 | 77 | 101 | ||
| BC058218.1 | - | 1 - 2 | 90 | 101 | ||
| BC092046.1 | - | 1 - 2 | 72 | 3594705 | 101 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK085754.1,AK142599.1,AK146549.1,AK149559.1,AK149595.1,AK168419.1,AK168938.1,BC012313.1,BC022986.1,BC092046.1,NM_133977.2 transcripts.
Set Membership: |
|

Trf
Genomic Map