Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 2 - 3 | 284 | 66 | |
| GenBank mRNA | AF440692.1 | - | 1 - 2 | 214 | 66 | |
| AK085754.1 | - | 2 - 3 | 269 | 66 | ||
| AK142599.1 | - | 274 | 66 | |||
| AK146549.1 | - | 2 - 3 | 269 | 66 | ||
| AK149559.1 | - | 2 - 3 | 269 | 66 | ||
| AK149595.1 | - | 2 - 3 | 269 | 66 | ||
| AK150782.1 | - | 2 - 3 | 317 | 66 | ||
| AK168368.1 | - | 2 - 3 | 269 | 66 | ||
| AK168405.1 | - | 2 - 3 | 269 | 66 | ||
| AK168419.1 | - | 2 - 3 | 269 | 66 | ||
| AK168938.1 | - | 2 - 3 | 269 | 66 | ||
| AK169187.1 | - | 2 - 3 | 269 | 66 | ||
| BC012313.1 | - | 2 - 3 | 284 | 3981648 | 66 | |
| BC020295.1 | - | 2 - 3 | 253 | 66 | ||
| BC022986.1 | - | 2 - 3 | 262 | 5123741 | 66 | |
| BC058216.1 | - | 2 - 3 | 250 | 66 | ||
| BC058218.1 | - | 2 - 3 | 263 | 66 | ||
| BC092046.1 | - | 2 - 3 | 245 | 3594705 | 66 | |
More Information
Set Membership: |
|

Trf
Genomic Map