Assay Details
| Gene Symbol: Trf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133977.2 | NP_598738.1 | 12 - 13 | 1557 | 68 | |
| GenBank mRNA | AF440692.1 | - | 11 - 12 | 1487 | 68 | |
| AK085754.1 | - | 12 - 13 | 1542 | 68 | ||
| AK142599.1 | - | 1547 | 68 | |||
| AK146549.1 | - | 12 - 13 | 1542 | 68 | ||
| AK149559.1 | - | 12 - 13 | 1542 | 68 | ||
| AK149595.1 | - | 12 - 13 | 1542 | 68 | ||
| AK150782.1 | - | 12 - 13 | 1590 | 68 | ||
| AK168368.1 | - | 12 - 13 | 1544 | 68 | ||
| AK168405.1 | - | 12 - 13 | 1544 | 68 | ||
| AK168419.1 | - | 12 - 13 | 1542 | 68 | ||
| AK168938.1 | - | 12 - 13 | 1542 | 68 | ||
| AK169187.1 | - | 12 - 13 | 1541 | 68 | ||
| BC008559.1 | - | 10 - 11 | 1279 | 68 | ||
| BC012313.1 | - | 12 - 13 | 1557 | 3981648 | 68 | |
| BC018573.1 | - | 7 - 8 | 827 | 68 | ||
| BC020295.1 | - | 12 - 13 | 1526 | 68 | ||
| BC022986.1 | - | 12 - 13 | 1535 | 5123741 | 68 | |
| BC058216.1 | - | 12 - 13 | 1523 | 68 | ||
| BC058218.1 | - | 12 - 13 | 1536 | 68 | ||
| BC092046.1 | - | 12 - 13 | 1518 | 3594705 | 68 | |
| S67218.1 | - | 1 - 2 | 109 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trf
Genomic Map