Assay Details
| Gene Symbol: Dgkg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138650.2 | NP_619591.1 | 10 - 11 | 1363 | 76 | |
| GenBank mRNA | AK083258.1 | - | 10 - 11 | 1356 | 76 | |
| AK087972.1 | - | 10 - 11 | 1321 | 76 | ||
| AK220294.1 | - | 10 - 11 | 1305 | 76 | ||
| BC015278.1 | - | 9 - 10 | 936 | 4221290 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkg
Genomic Map