Assay Details
| Gene Symbol: Atp12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138652.2 | NP_619593.2 | 4 - 5 | 631 | 138 | |
| GenBank mRNA | BC109010.1 | - | 4 - 5 | 629 | 138 | |
| BC109011.1 | - | 4 - 5 | 631 | 40048684 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp12a
Genomic Map