Assay Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013684.3 | NP_038712.3 | 5 - 6 | 855 | 105 | |
| GenBank mRNA | AK085037.1 | - | 5 - 6 | 856 | 105 | |
| BC012685.1 | - | 7 - 8 | 1091 | 3498542 | 105 | |
| BC050136.1 | - | 5 - 6 | 806 | 30020603 | 105 | |
| D01034.1 | - | 4 - 5 | 672 | 105 | ||
| U63933.1 | - | 6 - 7 | 861 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbp
Genomic Map