Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 4 - 5 | 462 | 109 | |
| GenBank mRNA | AK132254.1 | - | 4 - 5 | 431 | 109 | |
| AY424276.1 | - | 4 - 5 | 430 | 109 | ||
| BC014773.1 | - | 4 - 5 | 463 | 4193957 | 109 | |
| BC057186.1 | - | 4 - 5 | 437 | 5097123 | 109 | |
| J04766.1 | - | 4 - 5 | 427 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map