Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 5 - 6 | 606 | 94 | |
| GenBank mRNA | AK132254.1 | - | 5 - 6 | 575 | 94 | |
| AY424276.1 | - | 5 - 6 | 574 | 94 | ||
| BC014773.1 | - | 5 - 6 | 607 | 4193957 | 94 | |
| BC057186.1 | - | 5 - 6 | 581 | 5097123 | 94 | |
| J04766.1 | - | 5 - 6 | 571 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map