Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 9 - 10 | 1158 | 79 | |
| GenBank mRNA | AY424276.1 | - | 9 - 10 | 1126 | 79 | |
| BC014773.1 | - | 9 - 10 | 1159 | 4193957 | 79 | |
| BC057186.1 | - | 9 - 10 | 1133 | 5097123 | 79 | |
| J04766.1 | - | 9 - 10 | 1123 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map