Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 14 - 15 | 1868 | 65 | |
| GenBank mRNA | AK132254.1 | - | 14 - 15 | 1837 | 65 | |
| BC014773.1 | - | 14 - 15 | 1869 | 4193957 | 65 | |
| BC057186.1 | - | 14 - 15 | 1843 | 5097123 | 65 | |
| J04766.1 | - | 14 - 15 | 1833 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map