Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 15 - 16 | 1942 | 93 | |
| GenBank mRNA | AK132254.1 | - | 15 - 16 | 1911 | 93 | |
| BC014773.1 | - | 15 - 16 | 1943 | 4193957 | 93 | |
| BC057186.1 | - | 15 - 16 | 1917 | 5097123 | 93 | |
| J04766.1 | - | 15 - 16 | 1907 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plg
Genomic Map