Assay Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008891.2 | NP_032917.2 | 3 - 4 | 344 | 77 | |
| GenBank mRNA | AK009055.1 | - | 3 - 4 | 299 | 77 | |
| AK028219.1 | - | 3 - 4 | 285 | 77 | ||
| AK030978.1 | - | 3 - 4 | 273 | 77 | ||
| AK050669.1 | - | 3 - 4 | 290 | 77 | ||
| AK051640.1 | - | 3 - 4 | 342 | 77 | ||
| AK157587.1 | - | 3 - 4 | 288 | 77 | ||
| AK160616.1 | - | 3 - 4 | 279 | 77 | ||
| AK161109.1 | - | 3 - 4 | 284 | 77 | ||
| BC132571.1 | - | 3 - 4 | 278 | 40130848 | 77 | |
| BC138014.1 | - | 3 - 4 | 278 | 8861033 | 77 | |
| Y08701.1 | - | 3 - 4 | 278 | 77 | ||
More Information
Set Membership: |
|

Pnn
Genomic Map