Assay Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008959.3 | NP_032985.2 | 4 - 5 | 1118 | 77 | |
| GenBank mRNA | AF042731.1 | - | 4 - 5 | 533 | 77 | |
| AK036990.1 | - | 4 - 5 | 580 | 77 | ||
| AK054101.1 | - | 4 - 5 | 557 | 77 | ||
| AK085857.1 | - | 4 - 5 | 1119 | 77 | ||
| AK086136.1 | - | 3 - 4 | 642 | 77 | ||
| AK088048.1 | - | 4 - 5 | 600 | 77 | ||
| AK137681.1 | - | 4 - 5 | 788 | 77 | ||
| AK172873.1 | - | 4 - 5 | 1095 | 77 | ||
| BC003260.1 | - | 4 - 5 | 755 | 3501141 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptdss1
Genomic Map