Assay Details
| Gene Symbol: Sla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029841.4 | NP_001025012.1 | 6 - 7 | 781 | 58 | |
| NM_009192.3 | NP_033218.1 | 7 - 8 | 1050 | 58 | ||
| GenBank mRNA | AJ131777.1 | - | 6 - 7 | 688 | 58 | |
| AK036167.1 | - | 6 - 7 | 782 | 58 | ||
| AK041565.1 | - | 5 - 6 | 560 | 58 | ||
| AK137954.1 | - | 6 - 7 | 716 | 58 | ||
| BC032922.1 | - | 6 - 7 | 586 | 1430090 | 58 | |
| CT010282.1 | - | 5 - 6 | 484 | 58 | ||
| U29056.1 | - | 5 - 6 | 502 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sla
Genomic Map