Assay Details
| Gene Symbol: Snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042451.2 | NP_001035916.1 | 2 - 3 | 430 | 71 | |
| NM_009221.2 | NP_033247.1 | 2 - 3 | 334 | 71 | ||
| GenBank mRNA | AF033261.1 | - | 2 - 3 | 257 | 71 | |
| AF044672.1 | - | 1 - 2 | 162 | 71 | ||
| AF179273.1 | - | 2 - 3 | 334 | 71 | ||
| AK156316.1 | - | 2 - 3 | 266 | 71 | ||
| AK205246.1 | - | 1 - 2 | 156 | 71 | ||
| BC046764.1 | - | 2 - 3 | 173 | 5693785 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snca
Genomic Map