Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110145.1 | NP_001103615.1 | 4 - 5 | 524 | 116 | |
| NM_009278.4 | NP_033304.1 | 4 - 5 | 551 | 116 | ||
| GenBank mRNA | AK017822.1 | - | 4 - 5 | 465 | 116 | |
| AK032940.1 | - | 4 - 5 | 468 | 116 | ||
| AK088650.1 | - | 4 - 5 | 507 | 116 | ||
| AK088677.1 | - | 4 - 5 | 458 | 116 | ||
| AK134864.1 | - | 4 - 5 | 489 | 116 | ||
| AK141487.1 | - | 4 - 5 | 446 | 116 | ||
| BC003820.1 | - | 4 - 5 | 411 | 3593742 | 116 | |
| BC106150.1 | - | 4 - 5 | 404 | 3483153 | 116 | |
| L00993.1 | - | 4 - 5 | 487 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssb
Genomic Map