Assay Details
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110145.1 | NP_001103615.1 | 10 - 11 | 1182 | 91 | |
| NM_009278.4 | NP_033304.1 | 10 - 11 | 1209 | 91 | ||
| GenBank mRNA | AK017822.1 | - | 9 - 10 | 1021 | 91 | |
| AK032940.1 | - | 10 - 11 | 1126 | 91 | ||
| AK088650.1 | - | 10 - 11 | 1165 | 91 | ||
| AK088677.1 | - | 10 - 11 | 1116 | 91 | ||
| AK141487.1 | - | 10 - 11 | 1104 | 91 | ||
| AK168828.1 | - | 10 - 11 | 1132 | 91 | ||
| BC003820.1 | - | 10 - 11 | 1069 | 3593742 | 91 | |
| BC106150.1 | - | 10 - 11 | 1062 | 3483153 | 91 | |
| L00993.1 | - | 10 - 11 | 1145 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssb
Genomic Map