Assay Details
| Gene Symbol: Hnf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291268.1 | NP_001278197.1 | 1 - 2 | 529 | 114 | |
| NM_009330.3 | NP_033356.2 | 1 - 2 | 529 | 114 | ||
| GenBank mRNA | AB008174.1 | - | 1 - 2 | 349 | 114 | |
| AB008175.1 | - | 1 - 2 | 349 | 114 | ||
| AB008176.1 | - | 1 - 2 | 349 | 114 | ||
| AB008177.1 | - | 1 - 2 | 349 | 114 | ||
| AB052659.1 | - | 1 - 2 | 349 | 114 | ||
| AK004837.2 | - | 1 - 2 | 530 | 114 | ||
| AK019258.1 | - | 1 - 2 | 161 | 114 | ||
| X55842.1 | - | 1 - 2 | 349 | 114 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK019258.1 transcripts.
Set Membership: |
|

Hnf1b
Genomic Map