Assay Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289550.1 | NP_001276479.1 | 6 - 7 | 700 | 95 | |
| NM_001289551.1 | NP_001276480.1 | 6 - 7 | 700 | 95 | ||
| NM_001289552.1 | NP_001276481.1 | 5 - 6 | 557 | 95 | ||
| NM_001289553.1 | NP_001276482.1 | 5 - 6 | 584 | 95 | ||
| GenBank mRNA | AK003170.1 | - | 6 - 7 | 535 | 95 | |
| AK041233.1 | - | 6 - 7 | 528 | 96 | ||
| AK085042.1 | - | 5 - 6 | 426 | 95 | ||
| AK087114.1 | - | 6 - 7 | 535 | 95 | ||
| AK156423.1 | - | 6 - 7 | 535 | 95 | ||
| AK158409.1 | - | 5 - 6 | 409 | 95 | ||
| BC002049.1 | - | 6 - 7 | 534 | 3489459 | 95 | |
| BC056362.1 | - | 5 - 6 | 1809 | 6816930 | 95 | |
| DQ143891.1 | - | 5 - 6 | 585 | 95 | ||
| DQ143892.1 | - | 4 - 5 | 371 | 95 | ||
| DQ143893.1 | - | 4 - 5 | 378 | 95 | ||
| DQ143894.1 | - | 4 - 5 | 373 | 95 | ||
| DQ143895.1 | - | 4 - 5 | 789 | 95 | ||
| DQ143896.1 | - | 4 - 5 | 750 | 95 | ||
| DQ143897.1 | - | 4 - 5 | 828 | 95 | ||
| DQ143898.1 | - | 4 - 5 | 777 | 95 | ||
| DQ143899.1 | - | 3 - 4 | 904 | 95 | ||
| DQ143900.1 | - | 5 - 6 | 597 | 95 | ||
| L22482.1 | - | 5 - 6 | 661 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfb1i1
Genomic Map