Assay Details
| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009377.1 | NP_033403.1 | 1 - 2 | 100 | 65 | |
| GenBank mRNA | M69200.1 | - | 1 - 2 | 100 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Th
Genomic Map