Assay Details
| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009377.1 | NP_033403.1 | 6 - 7 | 702 | 62 | |
| GenBank mRNA | AK139568.1 | - | 919 | 62 | ||
| M69200.1 | - | 6 - 7 | 702 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Th
Genomic Map