Assay Details
| Gene Symbol: Th | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009377.1 | NP_033403.1 | 11 - 12 | 1204 | 57 | |
| GenBank mRNA | AK139568.1 | - | 1421 | 57 | ||
| M69200.1 | - | 11 - 12 | 1204 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Th
Genomic Map