Assay Details
| Gene Symbol: Top3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009410.2 | NP_033436.1 | 15 - 16 | 2142 | 113 | |
| GenBank mRNA | AB006074.1 | - | 15 - 16 | 2143 | 113 | |
| AK029987.1 | - | 12 - 13 | 1535 | 113 | ||
| AK037640.1 | - | 15 - 16 | 2043 | 113 | ||
| AK164443.1 | - | 15 - 16 | 2043 | 113 | ||
| AK170291.1 | - | 16 - 17 | 1949 | 113 | ||
| AK171286.1 | - | 16 - 17 | 1949 | 113 | ||
| BC099689.1 | - | 15 - 16 | 1957 | 30731611 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3a
Genomic Map