Assay Details
| Gene Symbol: Top3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009410.2 | NP_033436.1 | 17 - 18 | 2405 | 82 | |
| GenBank mRNA | AB006074.1 | - | 17 - 18 | 2406 | 82 | |
| AK029987.1 | - | 14 - 15 | 1798 | 82 | ||
| AK037640.1 | - | 17 - 18 | 2306 | 82 | ||
| AK170291.1 | - | 18 - 19 | 2212 | 82 | ||
| AK171286.1 | - | 18 - 19 | 2212 | 82 | ||
| BC099689.1 | - | 17 - 18 | 2220 | 30731611 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3a
Genomic Map