Assay Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159401.1 | NP_001152873.1 | 2 - 3 | 227 | 77 | |
| NM_001159402.1 | NP_001152874.1 | 2 - 3 | 226 | 77 | ||
| NM_009477.3 | NP_033503.2 | 2 - 3 | 254 | 77 | ||
| GenBank mRNA | AK136474.1 | - | 2 - 3 | 107 | 77 | |
| AL118479.1 | - | 2 - 3 | 227 | 77 | ||
| AV447018.1 | - | 2 - 3 | 278 | 77 | ||
| BC036559.1 | - | 2 - 3 | 128 | 5144694 | 77 | |
| CA493594.1 | - | 264 | 77 | |||
| CA536983.1 | - | 2 - 3 | 231 | 77 | ||
| D44464.1 | - | 2 - 3 | 255 | 77 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK136474.1,BC036559.1,D44464.1,NM_001159401.1,NM_001159402.1,NM_009477.3 transcripts.
Set Membership: |
|

Upp1
Genomic Map