Assay Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159401.1 | NP_001152873.1 | 7 - 8 | 906 | 55 | |
| NM_001159402.1 | NP_001152874.1 | 7 - 8 | 905 | 55 | ||
| NM_009477.3 | NP_033503.2 | 7 - 8 | 933 | 55 | ||
| GenBank mRNA | AK136474.1 | - | 7 - 8 | 786 | 55 | |
| BC036559.1 | - | 7 - 8 | 807 | 5144694 | 55 | |
| BX527866.1 | - | 1 - 2 | 150 | 55 | ||
| D44464.1 | - | 7 - 8 | 934 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upp1
Genomic Map