Assay Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145902.1 | - | 1 - 2 | 137 | 76 | |
| U16216.1 | - | 1 - 2 | 142 | 76 | ||
| U18867.2 | - | 1 - 2 | 146 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145902.1, U16216.1, U18867.2 transcripts.
Set Membership: |
|

Uros
Genomic Map