| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700037215.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037215.mm.1 |
TC0700004428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037205.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037205.mm.1 |
TC0700004428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037210.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037210.mm.1 |
TC0700004428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037206.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037206.mm.1 |
TC0700004428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037208.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037208.mm.1 |
TC0700004428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700037209.mm.1 |
NM_009479.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700037209.mm.1 |
TC0700004428.mm.1 |
MTA TC |