Assay Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA272198.1 | - | 3 - 4 | 351 | 72 | |
| AK135314.1 | - | 4 - 5 | 407 | 72 | ||
| AK145902.1 | - | 4 - 5 | 457 | 72 | ||
| AK148116.1 | - | 4 - 5 | 594 | 72 | ||
| AK164297.1 | - | 4 - 5 | 372 | 72 | ||
| BC068161.1 | - | 4 - 5 | 400 | 30358504 | 72 | |
| U04439.1 | - | 3 - 4 | 321 | 72 | ||
| U16216.1 | - | 4 - 5 | 462 | 72 | ||
| U18867.2 | - | 4 - 5 | 466 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uros
Genomic Map