Assay Details
| Gene Symbol: Pcyt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009981.4 | NP_034111.1 | 1 - 2 | 103 | 96 | |
| GenBank mRNA | AK044585.1 | - | 1 - 2 | 104 | 96 | |
| BB642590.1 | - | 1 - 2 | 104 | 96 | ||
| L28956.1 | - | 1 - 2 | 72 | 95 | ||
| Z12302.1 | - | 1 - 2 | 62 | 94 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the L28956.1,NM_009981.4,Z12302.1 transcripts.
Set Membership: |
|

Pcyt1a
Genomic Map