Assay Details
| Gene Symbol: Sparcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010097.4 | NP_034227.3 | 7 - 8 | 2215 | 76 | |
| GenBank mRNA | AK134608.1 | - | 7 - 8 | 1856 | 76 | |
| AK158725.1 | - | 7 - 8 | 1722 | 76 | ||
| BC003759.1 | - | 7 - 8 | 1860 | 3486162 | 76 | |
| U64827.1 | - | 7 - 8 | 1813 | 76 | ||
| U77330.1 | - | 7 - 8 | 1880 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sparcl1
Genomic Map