Assay Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010833.2 | NP_034963.2 | 5 - 6 | 739 | 84 | |
| GenBank mRNA | AK088336.1 | - | 5 - 6 | 687 | 84 | |
| AK133819.1 | - | 5 - 6 | 743 | 84 | ||
| AK145746.1 | - | 5 - 6 | 743 | 84 | ||
| AK150869.1 | - | 5 - 6 | 742 | 84 | ||
| AK153210.1 | - | 5 - 6 | 742 | 84 | ||
| AK157701.1 | - | 5 - 6 | 694 | 84 | ||
| BC003474.1 | - | 5 - 6 | 730 | 84 | ||
| BC047366.1 | - | 5 - 6 | 733 | 3711212 | 84 | |
| CF584663.1 | - | 227 | 84 | |||
| M86390.1 | - | 4 - 5 | 84 | |||
| S47577.1 | - | 5 - 6 | 724 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msn
Genomic Map