Assay Details
| Gene Symbol: Ncoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010881.2 | NP_035011.1 | 13 - 14 | 3327 | 98 | |
| GenBank mRNA | BC068177.1 | - | 10 - 11 | 2635 | 30628581 | 98 |
| BC080866.1 | - | 13 - 14 | 3090 | 30532122 | 98 | |
| U56920.1 | - | 12 - 13 | 2885 | 98 | ||
| U64606.1 | - | 10 - 11 | 2630 | 98 | ||
| U64828.1 | - | 13 - 14 | 3240 | 98 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089302.1 | - | Chr. - - [-] on Build | 98 | |||
More Information
Set Membership: |
|

Ncoa1
Genomic Map