Assay Details
| Gene Symbol: Nfia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122952.1 | NP_001116424.1 | 3 - 4 | 732 | 82 | |
| NM_001122953.1 | NP_001116425.1 | 3 - 4 | 971 | 82 | ||
| NM_010905.3 | NP_035035.1 | 3 - 4 | 971 | 82 | ||
| GenBank mRNA | AF326553.1 | - | 3 - 4 | 670 | 82 | |
| AF326554.1 | - | 3 - 4 | 670 | 82 | ||
| AK050320.1 | - | 2 - 3 | 288 | 82 | ||
| AK052204.1 | - | 3 - 4 | 793 | 82 | ||
| AK054310.1 | - | 3 - 4 | 835 | 82 | ||
| AK137731.1 | - | 3 - 4 | 834 | 82 | ||
| AK139357.1 | - | 3 - 4 | 733 | 82 | ||
| AK173173.1 | - | 2 - 3 | 607 | 82 | ||
| BC075702.1 | - | 3 - 4 | 952 | 30087137 | 82 | |
| BC122878.1 | - | 3 - 4 | 693 | 40107695 | 82 | |
| BC122880.1 | - | 3 - 4 | 691 | 82 | ||
| D90172.1 | - | 3 - 4 | 901 | 82 | ||
| D90173.1 | - | 3 - 4 | 809 | 82 | ||
| D90174.1 | - | 3 - 4 | 842 | 82 | ||
| D90175.1 | - | 686 | 82 | |||
| D90176.1 | - | 3 - 4 | 724 | 82 | ||
| U57633.1 | - | 3 - 4 | 626 | 82 | ||
| Y07690.1 | - | 3 - 4 | 702 | 82 | ||
| Y07691.1 | - | 3 - 4 | 700 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfia
Genomic Map