Assay Details
| Gene Symbol: Nrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164226.1 | NP_001157698.1 | 3 - 4 | 518 | 63 | |
| NM_001164227.1 | NP_001157699.1 | 3 - 4 | 518 | 63 | ||
| NM_001164228.1 | NP_001157700.1 | 3 - 4 | 515 | 63 | ||
| NM_001164230.1 | NP_001157702.1 | 3 - 4 | 515 | 63 | ||
| NM_010938.4 | NP_035068.3 | 3 - 4 | 515 | 63 | ||
| GenBank mRNA | AF098077.1 | - | 3 - 4 | 450 | 63 | |
| AK014494.1 | - | 3 - 4 | 479 | 63 | ||
| AK029034.1 | - | 3 - 4 | 496 | 63 | ||
| AK036604.1 | - | 3 - 4 | 476 | 63 | ||
| AK037697.1 | - | 3 - 4 | 476 | 63 | ||
| AK050861.1 | - | 3 - 4 | 432 | 63 | ||
| AK082580.1 | - | 3 - 4 | 516 | 63 | ||
| AK134483.1 | - | 3 - 4 | 432 | 63 | ||
| BC005410.1 | - | 2 - 3 | 336 | 3496661 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrf1
Genomic Map