Assay Details
| Gene Symbol: Sqstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290769.1 | NP_001277698.1 | 3 - 4 | 594 | 87 | |
| NM_011018.3 | NP_035148.1 | 3 - 4 | 594 | 87 | ||
| GenBank mRNA | BC006019.1 | - | 3 - 4 | 591 | 3487289 | 87 |
| E08146.1 | - | 3 - 4 | 552 | 87 | ||
| U40930.1 | - | 3 - 4 | 564 | 87 | ||
| U57413.1 | - | 3 - 4 | 556 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqstm1
Genomic Map