Assay Details
| Gene Symbol: Sqstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290769.1 | NP_001277698.1 | 4 - 5 | 734 | 76 | |
| NM_011018.3 | NP_035148.1 | 4 - 5 | 734 | 76 | ||
| GenBank mRNA | BC006019.1 | - | 4 - 5 | 731 | 3487289 | 76 |
| E08146.1 | - | 4 - 5 | 692 | 76 | ||
| U40930.1 | - | 4 - 5 | 704 | 76 | ||
| U57413.1 | - | 4 - 5 | 696 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sqstm1
Genomic Map