Assay Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082531.1 | NP_001076000.1 | 2 - 3 | 75 | 73 | |
| NR_002926.3 | 2 - 3 | 94 | 73 | |||
| GenBank mRNA | AK131857.1 | - | 2 - 3 | 95 | 73 | |
| BC045156.1 | - | 2 - 3 | 75 | 6479080 | 73 | |
| U32359.1 | - | 1 - 2 | 49 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2a
Genomic Map