Assay Details
| Gene Symbol: Polb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011130.2 | NP_035260.1 | 12 - 13 | 833 | 130 | |
| GenBank mRNA | AF513911.1 | - | 12 - 13 | 810 | 130 | |
| AK077127.1 | - | 12 - 13 | 818 | 130 | ||
| AK146745.1 | - | 12 - 13 | 822 | 130 | ||
| AK151436.1 | - | 12 - 13 | 818 | 130 | ||
| BC060998.1 | - | 12 - 13 | 814 | 6774075 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polb
Genomic Map