Assay Details
| Gene Symbol: Ppef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011148.3 | NP_035278.1 | 11 - 12 | 1523 | 91 | |
| GenBank mRNA | AF023458.1 | - | 11 - 12 | 1523 | 91 | |
| BC027049.1 | - | 12 - 13 | 1830 | 5356685 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppef2
Genomic Map