Assay Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011212.3 | NP_035342.3 | 15 - 16 | 1711 | 64 | |
| GenBank mRNA | AK004602.1 | - | 15 - 16 | 1713 | 64 | |
| AK132455.1 | - | 14 - 15 | 1674 | 64 | ||
| AK154910.1 | - | 12 - 13 | 1263 | 64 | ||
| BC053408.1 | - | 15 - 16 | 1487 | 64 | ||
| BC098473.1 | - | 15 - 16 | 1475 | 64 | ||
| D83484.1 | - | 15 - 16 | 2048 | 64 | ||
| U35368.1 | - | 15 - 16 | 1711 | 64 | ||
| U36758.1 | - | 12 - 13 | 1260 | 64 | ||
| U40280.1 | - | 14 - 15 | 1451 | 64 | ||
| U62387.1 | - | 15 - 16 | 1613 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpre
Genomic Map