Assay Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011223.3 | NP_035353.1 | 4 - 5 | 675 | 150 | |
| NM_133915.3 | NP_598676.2 | 4 - 5 | 675 | 150 | ||
| GenBank mRNA | AF293882.1 | - | 4 - 5 | 600 | 150 | |
| AF293883.1 | - | 3 - 4 | 495 | 150 | ||
| AK084243.1 | - | 2 - 3 | 340 | 150 | ||
| AK148060.1 | - | 4 - 5 | 661 | 150 | ||
| AK149933.1 | - | 4 - 5 | 640 | 150 | ||
| AK167299.1 | - | 4 - 5 | 614 | 150 | ||
| AK171436.1 | - | 4 - 5 | 640 | 150 | ||
| BC025493.1 | - | 4 - 5 | 633 | 150 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089534.1 | - | Chr. - - [-] on Build | 150 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pxn
Genomic Map